1-Bromo-2-((4-fluorobenzyl)oxy)benzene 97%

Supplier: Ambeed
Warning

A420138-100MG A420138-1G A420138-250MG A420138-5G
77196-214EA 36.05 USD
77196-214 77196-216 77196-218 77196-220
1-Bromo-2-((4-fluorobenzyl)oxy)benzene 97%
1-Bromo-2-((4-fluorobenzyl)oxy)benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
MDL Number: MFCD07782686
CAS Number: 494772-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR