(S)-3-Amino-3-cyclohexylpropanoic acid 95%

Supplier: Ambeed
Warning

A419986-100MG A419986-1G A419986-250MG
77168-522EA 9 USD
77168-522 77168-524 77892-865
(S)-3-Amino-3-cyclohexylpropanoic acid 95%
(S)-3-Amino-3-cyclohexylpropanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MDL Number: MFCD07373036
CAS Number: 91383-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR