(1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile monohydrate ≥99%

Supplier: Ambeed
Warning

Synonyms: Saxagliptin hydrate

A419052-10MG A419052-1MG A419052-25MG A419052-50MG A419052-5MG
77168-502EA 9.27 USD
77168-502 77168-504 77168-506 77168-508 77168-510
(1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile monohydrate ≥99%
(1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile monohydrate

Precaución: for Research Use Only

Formula: C₁₈H₂₇N₃O₃
MW: 333.43 g/mol
MDL Number: MFCD26131254
CAS Number: 945667-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR