2,4-Dichloro-6-iodophenol 97%

Supplier: Ambeed
Warning

A417932-100MG A417932-1G A417932-250MG A417932-5G
77196-178EA 29.87 USD
77196-178 77196-180 77196-182 77196-184
2,4-Dichloro-6-iodophenol 97%
2,4-Dichloro-6-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂IO
MW: 288.89 g/mol
MDL Number: MFCD00086134
CAS Number: 2040-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR