1,3,5-Triisopropylbenzene 95%

Supplier: Ambeed
Warning

A417498-100G A417498-25G A417498-500G
77196-164EA 33 USD
77196-164 77196-166 77196-168
1,3,5-Triisopropylbenzene 95%
1,3,5-Triisopropylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₂₄
MW: 204.36 g/mol
Boiling Pt: 232
Melting Pt: -7
MDL Number: MFCD00008890
CAS Number: 717-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR