1-Boc-5-chloroindole-2-boronic acid 98%

Supplier: Ambeed
Warning

A417465-1G A417465-250MG
77168-482EA 75 USD
77168-482 77168-484
1-Boc-5-chloroindole-2-boronic acid 98%
1-Boc-5-chloroindole-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅BClNO₄
MDL Number: MFCD04039005
CAS Number: 475102-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR