Ethyl 6-hydroxypyridazine-3-carboxylate 98%

Supplier: Ambeed
Warning

A417204-1G A417204-250MG
77196-160EA 28 USD
77196-160 77196-162
Ethyl 6-hydroxypyridazine-3-carboxylate 98%
Ethyl 6-hydroxypyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD11044476
CAS Number: 63001-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR