2-Aminobutanamide hydrochloride 95%

Supplier: Ambeed
Warning

A417132-1G A417132-25G A417132-5G
77168-476EA 13.39 USD
77168-476 77168-478 77168-480
2-Aminobutanamide hydrochloride 95%
2-Aminobutanamide hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂O
MW: 138.59 g/mol
MDL Number: MFCD03939986
CAS Number: 89603-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR