(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-fluorotetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 98%

Supplier: Ambeed
Danger

Synonyms: 2'-F-Bz-dC Phosphoramidite

A416844-1G A416844-250MG A416844-5G
77168-472EA 129.78 USD
77168-472 77613-278 77684-169
(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-fluorotetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 98%
(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-fluorotetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite

Precaución: For research use only.

Formula: C₄₆H₅₁FN₅O₈P
MW: 851.89 g/mol
MDL Number: MFCD18910581
CAS Number: 161442-19-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR