1,6-Bis(acryloyloxy)hexane 90% stabilised

Supplier: Ambeed
Warning

Synonyms: HDODA, Hexamethylene Glycol Diacrylate, Hexamethylene diacrylate, 1,6-Hexanediol Diacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A415936-100G A415936-500G A415936-25G
77196-156EA 15 USD
77196-156 77196-158 77594-288
1,6-Bis(acryloyloxy)hexane 90% stabilised
1,6-Bis(acryloyloxy)hexane

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₄
MDL Number: MFCD00008631
CAS Number: 13048-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR