2,6-Dichloro-1,7-naphthyridine 97%

Supplier: Ambeed
Warning

A414791-100MG A414791-1G A414791-250MG
77196-148EA 108.15 USD
77196-148 77196-150 77196-152
2,6-Dichloro-1,7-naphthyridine 97%
2,6-Dichloro-1,7-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MW: 199.03 g/mol
MDL Number: MFCD11044727
CAS Number: 93493-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR