2-Methyl-5-propionylfuran 97%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Methylfuran-2-yl)propan-1-one

A414078-1G A414078-25G A414078-5G
77196-134EA 6.18 USD
77196-134 77196-136 77196-138
2-Methyl-5-propionylfuran 97%
2-Methyl-5-propionylfuran

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00053142
CAS Number: 10599-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR