2-(2-Chloroethoxy)benzenesulfonamide 99%

Supplier: Ambeed
Warning

A413708-25G A413708-5G
77196-120EA 37 USD
77196-120 77196-122
2-(2-Chloroethoxy)benzenesulfonamide 99%
2-(2-Chloroethoxy)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₃S
MDL Number: MFCD03265372
CAS Number: 82097-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR