1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine 98%
Supplier: Ambeed
|
Warning
|
A413541-1G
A413541-25G
A413541-5G
77196-114EA
36
USD
77196-114
77196-116
77196-118
1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine 98%
1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine
Formula:
C₁₆H₂₄N₂O₂ |
MDL Number:
MFCD00059039 CAS Number: 99735-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...