1-Chloro-5-fluoroisoquinoline 96%

Supplier: Ambeed
Warning

A413086-100MG A413086-1G A413086-250MG
77168-460EA 133.9 USD
77168-460 77168-462 77168-464
1-Chloro-5-fluoroisoquinoline 96%
1-Chloro-5-fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClFN
MW: 181.59 g/mol
MDL Number: MFCD16883054
CAS Number: 435278-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR