1-(3-Chloro-4-methylphenyl)thiourea 95+%

Supplier: Ambeed
Danger

A413062-1G A413062-5G
77196-110EA 24.72 USD
77196-110 77196-112
1-(3-Chloro-4-methylphenyl)thiourea 95+%
1-(3-Chloro-4-methylphenyl)thiourea

Precaución: for Research Use Only

Formula: C₈H₉ClN₂S
MW: 200.6885 g/mol
MDL Number: MFCD00060443
CAS Number: 117174-84-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR