1-(4-Fluorophenyl)-1,3-butanedione 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluorobenzoyl)acetone, 1-(4-Fluorophenyl)butane-1,3-dione

A413033-1G A413033-25G A413033-5G
77196-104EA 30.9 USD
77196-104 77196-106 77196-108
1-(4-Fluorophenyl)-1,3-butanedione 98%
1-(4-Fluorophenyl)-1,3-butanedione

Precaución: for Research Use Only

Formula: C₁₀H₉FO₂
MW: 180.18 g/mol
MDL Number: MFCD00463139
CAS Number: 29681-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR