Decylamine 95%

Supplier: Ambeed
Danger

Synonyms: n-Decylamine, Decan-1-amine, 1-Aminodecane

A412856-10G A412856-1G A412856-25G A412856-5G
77196-090EA 32.96 USD
77196-090 77196-092 77196-094 77196-096
Decylamine 95%
Decylamine

Precaución: for Research Use Only

Formula: H₃C(CH₂)₈CH₂NH₂
MW: 157.3 g/mol
MDL Number: MFCD00008149
CAS Number: 2016-57-1
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR