1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methoxypyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxyuridine

A412544-100MG A412544-5G A412544-1G A412544-250MG
77168-458EA 22.66 USD
77168-458 77613-262 77613-258 77613-260
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methoxypyrimidine-2,4(1H,3H)-dione 95%
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methoxypyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₇
MW: 274.22 g/mol
MDL Number: MFCD00006533
CAS Number: 35542-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR