Methyl 4-bromo-3-formylbenzoate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A412138-100MG A412138-1G A412138-250MG A412138-5G A412138-25G
77168-450EA 15 USD
77168-450 77168-452 77168-454 77168-456 77684-157
Methyl 4-bromo-3-formylbenzoate 95%, Ambeed.Inc
Methyl 4-bromo-3-formylbenzoate

Precaución: For research use only.

Formula: C₉H₇BrO₃
MW: 243.054 g/mol
MDL Number: MFCD16036866
CAS Number: 858124-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR