2-Amino-N-methylthiazole-5-carboxamide 97%

Supplier: Ambeed
Warning

A411543-1G A411543-250MG
77168-438EA 255.44 USD
77168-438 77168-440
2-Amino-N-methylthiazole-5-carboxamide 97%
2-Amino-N-methylthiazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₅H₇N₃OS
MW: 157.19 g/mol
MDL Number: MFCD26770121
CAS Number: 1177494-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR