6,7-Dimethoxy-3,4-dihydroisoquinolin-1(2H)-one 96%

Supplier: Ambeed
Warning

A410514-100MG A410514-1G A410514-250MG A410514-5G
77168-432EA 39 USD
77168-432 77168-434 77168-436 77892-852
6,7-Dimethoxy-3,4-dihydroisoquinolin-1(2H)-one 96%
6,7-Dimethoxy-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD00197021
CAS Number: 493-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR