3,4-Dihydroxy-5-nitrobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A410286-1G
A410286-250MG
A410286-5G
A410286-25G
77168-426EA
29
USD
77168-426
77168-428
77168-430
77892-851
3,4-Dihydroxy-5-nitrobenzoic acid 95%
3,4-Dihydroxy-5-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅NO₆ |
MDL Number:
MFCD00872080 CAS Number: 84211-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...