(S)-2-Amino-3,3-dimethylbutanamide hydrochloride 98%

Supplier: Ambeed
Warning

A409928-100G A409928-1G A409928-250MG A409928-25G A409928-500G A409928-5G
77168-412EA 609.76 USD
77168-412 77168-414 77168-416 77168-418 77168-420 77168-422
(S)-2-Amino-3,3-dimethylbutanamide hydrochloride 98%
(S)-2-Amino-3,3-dimethylbutanamide hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₅ClN₂O
MW: 166.64 g/mol
MDL Number: MFCD21602493
CAS Number: 75158-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR