2',4'-Bis(benzyloxy)acetophenone 97%
Supplier: Ambeed
|
Warning
|
A409883-1G
A409883-250MG
A409883-5G
77168-408EA
48
USD
77168-408
77168-410
77892-850
2',4'-Bis(benzyloxy)acetophenone 97%
2',4'-Bis(benzyloxy)acetophenone
Precaución: for Research Use Only
Formula:
C₂₂H₂₀O₃ |
MDL Number:
MFCD00101153 CAS Number: 22877-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...