1-(4-Bromophenyl)-4,4,4-trifluorobutane-1,3-dione 97%

Supplier: Ambeed
Warning

A409278-1G A409278-5G
77196-058EA 28 USD
77196-058 77196-060
1-(4-Bromophenyl)-4,4,4-trifluorobutane-1,3-dione 97%
1-(4-Bromophenyl)-4,4,4-trifluorobutane-1,3-dione

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃O₂
MDL Number: MFCD00510742
CAS Number: 18931-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR