1,2,3,4,5,6-Hexahydropyrrolo[3,4-c]pyrrole dihydrobromide 95%

Supplier: Ambeed
Warning

A408756-100MG A408756-1G
77168-404EA 98.88 USD
77168-404 77168-406
1,2,3,4,5,6-Hexahydropyrrolo[3,4-c]pyrrole dihydrobromide 95%
1,2,3,4,5,6-Hexahydropyrrolo[3,4-c]pyrrole dihydrobromide

Precaución: for Research Use Only

Formula: C₆H₁₂Br₂N₂
MW: 271.98 g/mol
MDL Number: MFCD28405256
CAS Number: 135325-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR