Methyl 2-(dimethylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate 95+%

Supplier: Ambeed
Warning

A408686-100MG A408686-1G A408686-250MG
77168-398EA 36.05 USD
77168-398 77168-400 77168-402
Methyl 2-(dimethylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate 95+%
Methyl 2-(dimethylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate

Precaución: for Research Use Only

Formula: C₁₅H₂₃BN₂O₄
MW: 306.16 g/mol
MDL Number: MFCD16995283
CAS Number: 2304634-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR