2-(Hydroxymethyl)benzo[b]furan 95%

Supplier: Ambeed
Warning

Synonyms: Benzofuran-2-ylmethanol, (1-Benzofuran-2-yl)methanol, 2-(Hydroxymethyl)-1-benzofuran

A408404-100MG A408404-250MG
77196-054EA 20.6 USD
77196-054 77196-056
2-(Hydroxymethyl)benzo[b]furan 95%
2-(Hydroxymethyl)benzo[b]furan

Precaución: for Research Use Only

Formula: C₉H₈O₂
MW: 148.159 g/mol
MDL Number: MFCD00673995
CAS Number: 55038-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR