6-Bromo-4-chloroquinazoline 97%
Supplier: Ambeed
|
Warning
|
A408022-10G
A408022-1G
A408022-25G
77168-390EA
39.14
USD
77168-390
77168-392
77168-394
6-Bromo-4-chloroquinazoline 97%
6-Bromo-4-chloroquinazoline
Precaución: for Research Use Only
Formula:
C₈H₄BrClN₂ MW: 243.4878 g/mol |
MDL Number:
MFCD01862191 CAS Number: 38267-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...