Grazoprevir (MK-5172) 98%

Supplier: Ambeed
Warning

Synonyms: Grazoprevir, MK-5172, (33R,35S,91R,92R,5S)-5-(tert-butyl)-N-((1R,2S)-1-((cyclopropylsulphonyl)carbamoyl)-2-vinylcyclopropyl)-17-methoxy-4,7-dioxo-2,8-dioxa-6-aza-1(2,3)-quinoxalina-3(3,1)-pyrrolidina-9(1,2)-cyclopropanacyclotetradecaphane-35-carboxamide, Zepatier‎

A407625-10MG A407625-1MG A407625-25MG A407625-2MG A407625-50MG A407625-5MG
77168-378EA 329.6 USD
77168-378 77168-380 77168-382 77168-384 77168-386 77168-388
Grazoprevir (MK-5172) 98%
Grazoprevir (MK-5172)

Precaución: for Research Use Only

Formula: C₃₈H₅₀N₆O₉S
MW: 766.91 g/mol
MDL Number: MFCD22741510
CAS Number: 1350514-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR