2-Ethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A407547-10G A407547-1G A407547-25G A407547-5G A407547-100G
77196-038EA 29 USD
77196-038 77196-040 77196-042 77196-044 77746-820
2-Ethoxyphenylboronic acid 98%
2-Ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MDL Number: MFCD00674027
CAS Number: 213211-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR