6-Iodoimidazo[1,2-a]pyridine hydrochloride 97%

Supplier: Ambeed
Warning

A407421-100MG A407421-1G
77168-374EA 24.72 USD
77168-374 77168-376
6-Iodoimidazo[1,2-a]pyridine hydrochloride 97%
6-Iodoimidazo[1,2-a]pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₆ClIN₂
MW: 280.49 g/mol
MDL Number: MFCD28155366
CAS Number: 1205744-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR