(3S)-3-Aminoazepan-2-one 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Amino-hexahydro-2-azepinone, (S)-3-Amino-2-azepanone

A407373-1G A407373-250MG A407373-5G
77168-368EA 59.74 USD
77168-368 77168-370 77168-372
(3S)-3-Aminoazepan-2-one 98%
(3S)-3-Aminoazepan-2-one

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
MDL Number: MFCD00064476
CAS Number: 21568-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR