1,4-Dioxane-2,3-diol 96%

Supplier: Ambeed
Warning

A406492-1G A406492-5G
77196-024EA 13 USD
77196-024 77746-815
1,4-Dioxane-2,3-diol 96%
1,4-Dioxane-2,3-diol

Precaución: for Research Use Only

Formula: C₄H₈O₄
MDL Number: MFCD00006572
CAS Number: 4845-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR