3,5-Dichloro-2-fluorobenzaldehyde 95+%

Supplier: Ambeed
Danger

A406347-100MG A406347-1G A406347-250MG
77168-350EA 42.23 USD
77168-350 77168-352 77168-354
3,5-Dichloro-2-fluorobenzaldehyde 95+%
3,5-Dichloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
MDL Number: MFCD26687657
CAS Number: 477535-42-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR