o,α-Dimethylbenzyl alcohol 95%

Supplier: Ambeed
Warning

A405613-1G A405613-25G
77196-018EA 62.83 USD
77196-018 77196-020 77196-022
o,α-Dimethylbenzyl alcohol 95%
o,α-Dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00046632
CAS Number: 7287-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR