Thiobenzamide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Benzothioamide
A405532-100G
A405532-25G
A405532-5G
77196-006EA
46.35
USD
77196-006
77196-008
77196-010
Thiobenzamide 98%
Thiobenzamide
Precaución: for Research Use Only
Formula:
C₇H₇NS MW: 137.21 g/mol |
MDL Number:
MFCD00008060 CAS Number: 2227-79-4 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...