4-Bromo-5-(trifluoromethyl)thiophene-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A405122-100MG
A405122-250MG
A405122-1G
77168-322EA
111.24
USD
77168-322
77168-324
77568-822
4-Bromo-5-(trifluoromethyl)thiophene-2-carboxylic acid 95%
4-Bromo-5-(trifluoromethyl)thiophene-2-carboxylic acid
Precaución: For research use only.
Formula:
C₆H₂BrF₃O₂S MW: 275.04 g/mol |
MDL Number:
MFCD28099386 CAS Number: 1034188-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...