Pyrazolo[1,5-a]pyridine-7-carbaldehyde 98%

Supplier: Ambeed
Warning

A405060-250MG A405060-5G A405060-100MG
77168-320EA 36 USD
77168-320 77892-836 77892-835
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 98%
Pyrazolo[1,5-a]pyridine-7-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD06739029
CAS Number: 362661-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR