PF-06291874 98+%

Supplier: Ambeed
Warning

Synonyms: PF6291874., (S)-3-(4-(1-(3,5-Dimethyl-4-(4-(trifluoromethyl)-1H-pyrazol-1-yl)phenoxy)butyl)benzamido)propanoic acid, GCGR Antagonist IV, Glucagon receptor antagonists-4, N-[4-[(1S)-1-[3,5-Dimethyl-4-[4-(trifluoromethyl)-1H-pyrazol-1-yl]phenoxy]butyl]benzoyl]—alanine

A404945-10MG A404945-1MG A404945-25MG A404945-5MG
77195-984EA 73 USD
77195-984 77195-986 77195-988 77195-990
PF-06291874 98+%
PF-06291874

Precaución: for Research Use Only

Formula: C₂₆H₂₈F₃N₃O₄
MW: 503.51 g/mol
MDL Number: MFCD30343861
CAS Number: 1393124-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR