1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium chloride 97%
Supplier: Ambeed
|
Warning
|
A404755-10G
A404755-1G
A404755-250MG
A404755-5G
A404755-100G
A404755-25G
77195-976EA
114.33
USD
77195-976
77195-978
77195-980
77195-982
77746-811
77746-812
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium chloride 97%
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium chloride
Precaución: for Research Use Only
Formula:
C₂₁H₂₇ClN₂ MW: 342.91 g/mol |
MDL Number:
MFCD04973305 CAS Number: 173035-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...