tert-Butyl 6-formylindoline-1-carboxylate 95%

Supplier: Ambeed
Warning

A404579-100MG A404579-1G A404579-250MG
77168-314EA 55.62 USD
77168-314 77168-316 77168-318
tert-Butyl 6-formylindoline-1-carboxylate 95%
tert-Butyl 6-formylindoline-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD09878908
CAS Number: 391668-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR