1,2-Bis(dicyclohexylphosphino)benzene 98%

Supplier: Ambeed
Warning

A404222-100MG A404222-1G A404222-250MG A404222-5G
77195-964EA 92.7 USD
77195-964 77195-966 77195-968 77746-809
1,2-Bis(dicyclohexylphosphino)benzene 98%
1,2-Bis(dicyclohexylphosphino)benzene

Precaución: for Research Use Only

Formula: C₃₀H₄₈P₂
MW: 470.64 g/mol
MDL Number: MFCD01074565
CAS Number: 215951-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR