(R)-3-Amino-3-(thiophen-2-yl)propanoic acid 96%

Supplier: Ambeed
Warning

A403505-100MG A403505-250MG
77195-960EA 45.32 USD
77195-960 77195-962
(R)-3-Amino-3-(thiophen-2-yl)propanoic acid 96%
(R)-3-Amino-3-(thiophen-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
MDL Number: MFCD04113649
CAS Number: 73495-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR