(5-Fluorobenzofuran-2-yl)boronic acid 98%

Supplier: Ambeed
Warning

A401687-250MG A401687-1G A401687-100MG
77168-304EA 63 USD
77168-304 77572-246 77892-828
(5-Fluorobenzofuran-2-yl)boronic acid 98%
(5-Fluorobenzofuran-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₆BFO₃
MDL Number: MFCD18383019
CAS Number: 473416-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR