4-Cyano-3-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A397915-1G A397915-250MG A397915-5G
77168-272EA 248.23 USD
77168-272 77168-274 77168-276
4-Cyano-3-methoxybenzoic acid 95%
4-Cyano-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.15 g/mol
MDL Number: MFCD18203531
CAS Number: 102362-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR