3-Aminobenzamide 98%

Supplier: Ambeed
Warning

A397911-100MG A397911-250MG A397911-25G A397911-5G A397911-1G
77195-890EA 5 USD
77195-890 77195-892 77195-894 77195-896 77572-222
3-Aminobenzamide 98%
3-Aminobenzamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MDL Number: MFCD00007989
CAS Number: 3544-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR