N-(5-Iodothiazol-2-yl)acetamide 97%

Supplier: Ambeed
Warning

A396865-100MG A396865-1G A396865-250MG A396865-5G
77168-262EA 20.6 USD
77168-262 77168-264 77168-266 77168-268
N-(5-Iodothiazol-2-yl)acetamide 97%
N-(5-Iodothiazol-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₅H₅IN₂OS
MW: 268.08 g/mol
MDL Number: MFCD09701411
CAS Number: 252662-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR