AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
A395798-100MG
A395798-1MG
A395798-250MG
A395798-1G
77195-884EA
36
USD
77195-884
77195-886
77616-704
77746-795
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) 98%
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulphonylfluoride hydrochloride)
Precaución: for Research Use Only
Formula:
C₈H₁₁ClFNO₂S MW: 239.69 g/mol |
MDL Number:
MFCD00132962 CAS Number: 30827-99-7 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...